Imprimer

Anti-HB Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB24744-1EA 724 CHF
BIRBORB24744-1
Anti-HB Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: HB (hemoglobin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR