Imprimer

Anti-CK-MB Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB24715-1EA 0 CHF
BIRBORB24715-1
Anti-CK-MB Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: CK-MB
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR