Imprimer

Anti-Hemoglobin Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24649-1EA 743 CHF
BIRBORB24649-1
Anti-Hemoglobin Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Hemoglobin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR