Imprimer

Anti-APOA1 Goat Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB24576-1EA 595 CHF
BIRBORB24576-1
Anti-APOA1 Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR