Imprimer

Anti-Barbiturates (BAR) Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB24510-1EA 0 CHF
BIRBORB24510-1
Anti-Barbiturates (BAR) Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: Barbiturates (BAR)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR