Imprimer

Anti-Erythromycin Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24481-1EA 0 CHF
BIRBORB24481-1
Anti-Erythromycin Mouse Monoclonal Antibody
Anticorps
ELISA: 1:1500

Type: Primary
Antigen: Erythromycin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR