Imprimer

Anti-FKBP10 Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB24462-0.2EA 1226 CHF
BIRBORB24462-0.2
Anti-FKBP10 Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: FKBP10 (FK506 binding protein 10, 65 kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR