Imprimer

Anti-Oxycodone Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB24455-1EA 0 CHF
BIRBORB24455-1
Anti-Oxycodone Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Oxycodone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR