Imprimer

Anti-Vancomycin Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB24447-0.2EA 0 CHF
BIRBORB24447-0.2
Anti-Vancomycin Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Vancomycin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR