Imprimer

Anti-C-peptide (Proinsulin) Mouse Monoclonal Antibody [clone: 5B8]

Fournisseur: Biorbyt
BIRBORB24423-1EA 0 CHF
BIRBORB24423-1
Anti-C-peptide (Proinsulin) Mouse Monoclonal Antibody [clone: 5B8]
Anticorps
Type: Primary
Antigen: C-peptide (Proinsulin)
Clonality: monoclonal
Clone: 5B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR