Imprimer

Anti-PAPPA Mouse Monoclonal Antibody (Biotin)

Fournisseur: Biorbyt
BIRBORB24370-1EA 1571 CHF
BIRBORB24370-1
Anti-PAPPA Mouse Monoclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: PAPPA (pregnancy-associated plasma protein A, pappalysin 1)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR