Imprimer

Anti-Growth Hormone Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24369-1EA 615 CHF
BIRBORB24369-1
Anti-Growth Hormone Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Growth Hormone
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR