Imprimer

Anti-IGF1 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24338-0.2EA 914 CHF
BIRBORB24338-0.2
Anti-IGF1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: IGF1 (insulin-like growth factor 1 (somatomedin C))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR