Imprimer

Anti-Relaxin 40575 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24318-10EA 0 CHF
BIRBORB24318-10
Anti-Relaxin 40575 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Relaxin 40575
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR