Imprimer

Anti-TMSB10 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB24204-20EA 0 CHF
BIRBORB24204-20
Anti-TMSB10 Rabbit Polyclonal Antibody
Anticorps
ELISA: 1:4000

Type: Primary
Antigen: TMSB10 (thymosin beta 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR