Imprimer

Anti-Glucagon-Like Peptide 1 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24200-10EA 0 CHF
BIRBORB24200-10
Anti-Glucagon-Like Peptide 1 Mouse Monoclonal Antibody
Anticorps
  • Immunochemistry: 1:10000; DB: 1:5000

Type: Primary
Antigen: Glucagon-Like Peptide 1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR