Imprimer

Anti-Sialoprotein Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB24186-20EA 0 CHF
BIRBORB24186-20
Anti-Sialoprotein Rabbit Polyclonal Antibody
Anticorps
ELISA: 1:1000

Type: Primary
Antigen: Sialoprotein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR