Imprimer

Anti-HBcAg (ayw) Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB24149-100EA 0 CHF
BIRBORB24149-100
Anti-HBcAg (ayw) Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: HBcAg (ayw)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR