Imprimer

Anti-HPV 11 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24103-1EA 803 CHF
BIRBORB24103-1
Anti-HPV 11 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: HPV 11
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR