Imprimer

Anti-Hepatitis B 'e' Antigen Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24087-1EA 792 CHF
BIRBORB24087-1
Anti-Hepatitis B 'e' Antigen Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Hepatitis B 'e' Antigen
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR