Imprimer
Anti-Influenza B (NP) Mouse Monoclonal Antibody
Fournisseur: Biorbyt
BIRBORB24073-1EA 618 CHF
BIRBORB24073-1
Anti-Influenza B (NP) Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Influenza B (NP)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now