Imprimer
Anti-HBsAg (ayw) Mouse monoclonal antibody
Fournisseur: Biorbyt
BIRBORB24027-1EA 0 CHF
BIRBORB24027-1
Anti-HBsAg (ayw) Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: HBsAg (ayw)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now