Imprimer
Anti-Epstein Barr Virus Mouse Monoclonal Antibody
Fournisseur: Biorbyt
BIRBORB24026-1EA 0 CHF
BIRBORB24026-1
Anti-Epstein Barr Virus Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Epstein Barr Virus
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now