Imprimer

Anti-Fr. tularensis (LPS) Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24010-1EA 732 CHF
BIRBORB24010-1
Anti-Fr. tularensis (LPS) Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Fr. tularensis (LPS)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR