Imprimer

Anti-Staph. aureus Enterotoxin Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23994-1EA 587 CHF
BIRBORB23994-1
Anti-Staph. aureus Enterotoxin Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Staph. aureus Enterotoxin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR