Imprimer

Anti-Streptococcus, group B Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23881-100EA 476 CHF
BIRBORB23881-100
Anti-Streptococcus, group B Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Streptococcus, group B
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR