Imprimer

Anti-Neisseria gonorrhoeae Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23866-100EA 0 CHF
BIRBORB23866-100
Anti-Neisseria gonorrhoeae Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Neisseria gonorrhoeae
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR