Imprimer

Anti-BSA Chicken Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB23733-2EA 604 CHF
BIRBORB23733-2
Anti-BSA Chicken Polyclonal Antibody
Anticorps
Type: Primary
Antigen: BSA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: none
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR