Imprimer

Anti-Protein A Chicken Polyclonal Antibody (Biotin)

Fournisseur: Biorbyt
BIRBORB23732-2EA 1134 CHF
BIRBORB23732-2
Anti-Protein A Chicken Polyclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: Protein A
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Chicken
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR