Imprimer

Anti-Staphylococcus aureus Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23714-0.2EA 0 CHF
BIRBORB23714-0.2
Anti-Staphylococcus aureus Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Staphylococcus aureus
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR