Imprimer

Anti-C. trachomatis MOMP Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23678-1EA 961 CHF
BIRBORB23678-1
Anti-C. trachomatis MOMP Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: C. trachomatis MOMP
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR