Imprimer

Anti-AAV (intact) Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23666-50EA 0 CHF
BIRBORB23666-50
Anti-AAV (intact) Mouse Monoclonal Antibody
Anticorps
Immunofluorescence: 1:20; Immunoprecipitation: 1:5

Type: Primary
Antigen: AAV (intact)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR