Imprimer

Anti-Hepatitis B `e' Ag Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23660-1EA 1065 CHF
BIRBORB23660-1
Anti-Hepatitis B `e' Ag Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Hepatitis B `e' Ag
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR