Imprimer

Anti-Rotavirus, p43 (vp6) Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23638-1EA 606 CHF
BIRBORB23638-1
Anti-Rotavirus, p43 (vp6) Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Rotavirus, p43 (vp6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR