Imprimer

Anti-VZV (gpI + IV) Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23627-0.5EA 692 CHF
BIRBORB23627-0.5
Anti-VZV (gpI + IV) Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: VZV (gpI + IV)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR