Imprimer

Anti-HSV-1 (155kDa) Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23626-0.5EA 0 CHF
BIRBORB23626-0.5
Anti-HSV-1 (155kDa) Mouse Monoclonal Antibody
Anticorps
Immunofluorescence: 1:20-1:30

Type: Primary
Antigen: HSV-1 (155kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR