Imprimer

Anti-Adenovirus Hexon Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23619-1EA 0 CHF
BIRBORB23619-1
Anti-Adenovirus Hexon Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Adenovirus Hexon
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR