Imprimer

Anti-Japanese Encephalitis Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23558-100EA 0 CHF
BIRBORB23558-100
Anti-Japanese Encephalitis Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Japanese Encephalitis
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR