Imprimer

Anti-H. pylori CagA Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23539-1EA 777 CHF
BIRBORB23539-1
Anti-H. pylori CagA Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: H. pylori CagA
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR