Imprimer

Anti-HBsAg (ad/ay) Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB23483-1EA 749 CHF
BIRBORB23483-1
Anti-HBsAg (ad/ay) Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: HBsAg (ad/ay)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR