Imprimer
Anti-C. botulinum Toxin A Mouse Monoclonal Antibody
Fournisseur: Biorbyt
BIRBORB23430-1EA 0 CHF
BIRBORB23430-1
Anti-C. botulinum Toxin A Mouse Monoclonal Antibody
Anticorps
  • Immunofluorescence: 1:25
Type: Primary
Antigen: C. botulinum Toxin A
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now