Imprimer

Anti-Chlamydia species Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23425-100EA 476 CHF
BIRBORB23425-100
Anti-Chlamydia species Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Chlamydia species
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR