Imprimer

Anti-Rat EBV LMP2A Rat Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23362-0.25EA 0 CHF
BIRBORB23362-0.25
Anti-Rat EBV LMP2A Rat Monoclonal Antibody
Anticorps
Western Blot: 1:100-1:1000

Type: Primary
Antigen: Rat EBV LMP2A
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR