Imprimer

Anti-Marek Disease Virus Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB23343-1EA 1023 CHF
BIRBORB23343-1
Anti-Marek Disease Virus Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Marek Disease Virus
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR