Imprimer

Anti-ACE2 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB23290-100EA 0 CHF
BIRBORB23290-100
Anti-ACE2 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: ACE2 (angiotensin I converting enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR