Imprimer

Anti-RSV Goat Polyclonal Antibody (Biotin)

Fournisseur: Biorbyt
BIRBORB23241-1EA 677 CHF
BIRBORB23241-1
Anti-RSV Goat Polyclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: RSV
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR