Imprimer

Anti-HBsAg (ad/ay) Goat Polyclonal Antibody (Biotin)

Fournisseur: Biorbyt
BIRBORB23233-1EA 649 CHF
BIRBORB23233-1
Anti-HBsAg (ad/ay) Goat Polyclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: HBsAg (ad/ay)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR