Imprimer

Anti-Leptospira biflexa Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB23180-1EA 572 CHF
BIRBORB23180-1
Anti-Leptospira biflexa Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Leptospira biflexa
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR