Imprimer
Anti-Influenza B Virions Goat Polyclonal Antibody (Biotin)
Fournisseur: Biorbyt
BIRBORB23177-1EA 595 CHF
BIRBORB23177-1
Anti-Influenza B Virions Goat Polyclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: Influenza B Virions
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now