Imprimer

Anti-Influenza A Goat Polyclonal Antibody (FITC (Fluorescein))

Fournisseur: Biorbyt
BIRBORB23169-1EA 667 CHF
BIRBORB23169-1
Anti-Influenza A Goat Polyclonal Antibody (FITC (Fluorescein))
Anticorps
Type: Primary
Antigen: Influenza A
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR